Learn More
Azlon™ Polypropylene Measuring Scoop
Product Code. p-8001842
Click to view available options
Length (Metric):
60 mm
82 mm
102 mm
137 mm
160 mm
198 mm
259 mm
315 mm
385 mm
Capacity (Metric):
2 mL
5 mL
10 mL
25 mL
50 mL
100 mL
250 mL
500 mL
1000 mL
Unit Size:
Each
Pack of 5
Product Code. 11349633
Brand: Azlon SWL140
Scoop chemical reagents or food items safely. The Azlon™ polypropylene measuring scoop comes with flat top and bottom.
Scoop chemical reagents or food items safely. The Azlon™ polypropylene measuring scoop comes with flat top and bottom.
  • Made from food safe polypropylene
  • Top is flat for levelling off powders
  • Base is flat enabling scoops to stand horizontally
  • The capacity is clearly molded on each size
TRUSTED_SUSTAINABILITY
Specifications
Product Type Measuring Scoop
Material Polypropylene
Includes Handle
Capacity (Metric) 1000 mL
Sterility Non-sterile
Length (Metric) 385 mm