Learn More
Azlon™ Polyethylene Stoppers
Click to view available options
Standard Taper Joint:
10/19
12/21
14/23
19/26
24/29
29/32
34/35
Unit Size:
Pack of 10
Description
Use these stoppers with glass or plastic volumetric flasks and measuring cylinders which have longer 'B' length sockets. AzlonTM Polyethylene Stoppers feature a large head for a good grip and extra protection if glassware is knocked over.
- Manufactured from polyethylene
- Available in white and red
- Suitable for use with both Azlon and glass volumetric ware
- Enlarged head provides a good grip and protects glassware
- Ribbed cone ensures a secure leak free seal
Specifications
Specifications
| For Use With (Equipment) | Glass or Plastic volumetric flasks and measuring cylinders etc |
| Standard Taper Joint | 19/26 |
| Color | White, Red |
| Material | Polyethylene |
| Product Type | Tapered Stopper |
| Shape | Ribbed Cone |