Learn More
Azlon™ Micro Magnetic Stirrer Bar
Product Code. p-8001816
Click to view available options
Length (Metric):
7 mm
10 mm
13 mm
Unit Size:
Pack of 2
Product Code. 10359324
Brand: Azlon SWN954
Edge
Use these bars in for a variety of stirring applications. Fisherbrand™ Azlon™ Micro Magnetic Stirrer Bars are ideal for small vessels, test tubes and vials.
  • For efficient stirring
  • PTFE composition
TRUSTED_SUSTAINABILITY
Specifications
Autoclavable Autoclavable
Length (English) 0.3 in.
Diameter (Metric) 3 mm
Length (Metric) 10 mm
Material Magnet
Quantity 2/Pk.
Shape Micro