Learn More
Azlon™ Lightweight Polypropylene Funnels
Product Code. p-8001813
Click to view available options
Diameter (Metric) Stem:
5mm
9mm
10mm
13mm
14mm
17mm
Height (Metric):
47 mm
62 mm
78 mm
107 mm
149 mm
170 mm
210 mm
Unit Size:
Pack of 10
Pack of 5
Product Code. 12024608
Brand: Azlon FWC060
Lightweight polypropylene made from food grade plastic
Manufactured from lightweight polypropylene. Azlon™ Lightweight Polypropylene Funnels include ribs that prevent airlocks and allow rapid filtration. Autoclavable to 121 °C.
  • Lightweight polypropylene funnels with ribs that prevent air locks, allowing rapid filtration
  • Autoclavable to 121°C
  • Made from food grade plastic
TRUSTED_SUSTAINABILITY
Specifications
Diameter (Metric) Stem 10mm
Material Polypropylene
Height (Metric) 107 mm