Learn More
Azlon™ Lightweight Polypropylene Funnels
Product Code. 10708205
Click to view available options
Diameter (Metric) Stem:
5mm
9mm
10mm
13mm
14mm
17mm
Height (Metric):
47 mm
62 mm
78 mm
107 mm
149 mm
170 mm
210 mm
Unit Size:
Pack of 10
Pack of 5
Product Code. 10708205
Brand: Azlon FWC054
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
Lightweight polypropylene made from food grade plastic
Manufactured from lightweight polypropylene. Azlon™ Lightweight Polypropylene Funnels include ribs that prevent airlocks and allow rapid filtration. Autoclavable to 121 °C.
  • Lightweight polypropylene funnels with ribs that prevent air locks, allowing rapid filtration
  • Autoclavable to 121°C
  • Made from food grade plastic
TRUSTED_SUSTAINABILITY
Specifications
Diameter (Metric) Stem 5mm
Material Polypropylene
Height (Metric) 47 mm