Learn More
Azlon™ Polypropylene, Heavy Duty Aspirator Bottle
Click to view available options
:
Capacity: 20L
Description
- Raised graduations and translucent color supports visual volume assessment
- Heavy duty leak-proof stopcock opens and closes in a quarter turn
- Molded integral carry handle provides ease of transportation
- Autoclavable due to polypropylene composition
Specifications
Specifications
| Autoclavable | Autoclavable |
| Capacity (English) | 5.3 gal. |
| Capacity (Metric) | 20 L |
| Color | White (Translucent) |
| For Use With (Application) | For easy placement on shelves or bench top |
| Height (Metric) With Closure | 420 mm |
| Includes | Closure, Moulded carrying handle |
| Material | Polypropylene |
| Product Type | Aspirator Bottle |
| Shape | Octagonal |