Learn More
Azlon™ Cylindrical Stirrer Bar
Product Code. p-8001782
Click to view available options
Length (Metric):
6 mm
8 mm
12 mm
13 mm
15 mm
20 mm
25 mm
30 mm
35 mm
40 mm
45 mm
50 mm
60 mm
70 mm
80 mm
Diameter (Metric):
3 mm
4.5 mm
6 mm
8 mm
9 mm
9.5 mm
Unit Size:
Pack of 2
Product Code. 10606865
Brand: Azlon SWN652
Only null left
Add to basket
Add to basket
Available in a range of lengths. Azlon™ Cylindrical Stirrer Bar is suitable for a wide range of flat bottomed containers.
  • Plain design with rounded ends
  • Suitable for a wide range of flat bottom containers
TRUSTED_SUSTAINABILITY
Specifications
Autoclavable Autoclavable
Dimensions (L x W x H) 0.23 Lin.
Length (English) 0.23 in.
Coating PTFE
Diameter (Metric) 3 mm
Length (Metric) 6 mm
Material Magnet
Quantity 2/Pk.
Shape Cylinder
Type Stir Bar