Learn More
Azithromycin B, TRC
Product Code. 17954485
Change view
Click to view available options
Quantity:
1 mg
2.5 mg
10 mg
Unit Size:
10mg
1mg
2.50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17954485 10 mg 10mg
17954725 2.5 mg 2.50mg
17951725 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 17954485 Supplier Toronto Research Chemicals Supplier No. TRCA92700510MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 307974-61-4
Molecular Formula C38H72N2O11
Molecular Weight (g/mol) 732.99
Synonym 1-Oxa-6-azacyclopentadecan-15-one, 13-[(2,6-dideoxy-3-C-methyl-3-O-methyl-α-L-ribo-hexopyranosyl)oxy]-2-ethyl-4,10-dihydroxy-3,5,6,8,10,12,14-heptamethyl-11-[[3,4,6-trideoxy-3-(dimethylamino)-β-D-xylo-hexopyranosyl]oxy]-, (2R,3R,4R,5R,8R,10R,11R,12R,13S,14R)- (9CI), 3-Deoxyazithromycin, Azitromycin B, Azithromycin Imp. B (EP)
SMILES CC[C@H]1OC(=O)[C@H](C)[C@@H](O[C@H]2C[C@@](C)(OC)[C@@H](O)[C@H](C)O2)[C@H](C)[C@@H](O[C@@H]3O[C@H](C)C[C@@H]([C@H]3O)N(C)C)[C@](C)(O)C[C@@H](C)CN(C)[C@H](C)[C@@H](O)[C@H]1C
Specifications
Quantity 10 mg
Percent Purity >95%
Chemical Name or Material Azithromycin B
Grade HPLC