Learn More
Azidamfenicol, TRC
Specifications
Specifications
| Chemical Name or Material | Azidamfenicol |
| CAS | 13838-08-9 |
| Molecular Formula | C11H13N5O5 |
| Quantity | 5 mg |
| Synonym | 2-Azido-N-[(1R,2R)-2-hydroxy-1-(hydroxymethyl)-2-(4-nitrophenyl)ethyl]acetamide, Azidamfenicol, Azidamphenicol, Azidoamphenicol, D-(-)-threo-1-p-Nitrophenyl-2-azidoacetylamino-1,3-propanediol, Leukomycin N, D-Threo-(-)-2-azido-N-[beta-hydroxy-alpha-(hydroxymethyl)-p-nitrophenethyl]-acetamide, [R-(R*,R*)]-2-Azido-N-[2-hydroxy-1-(hydroxymethyl)-2-(4-nitrophenyl)ethyl]-acetamide |
| InChI Formula | InChI=1S/C11H13N5O5/c12-15-13-5-10(18)14-9(6-17)11(19)7-1-3-8(4-2-7)16(20)21/h1-4,9,11,17,19H,5-6H2,(H,14,18)/t9-,11-/m1/s1 |
| SMILES | OC[C@@H](NC(=O)CN=[N+]=[N-])[C@H](O)c1ccc(cc1)[N+](=O)[O-] |
| IUPAC Name | 2-azido-N-[(1R,2R)-1,3-dihydroxy-1-(4-nitrophenyl)propan-2-yl]acetamide |
| Molecular Weight (g/mol) | 295.25 |
| Formula Weight | 295.0917 |