missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ AZD 1480

Click to view available options
Quantity:
10 mg
Unit Size:
10mg
Description
AZD 1480 is a potent and selective JAK2 inhibitor (IC50 < 3 nM). Exhibits >50-fold selectivity for JAK2 over JAK3. Exhibits antitumor effects and inhibits formation of lung metastases in a mouse renal cancer model. Also inhibits STAT3 signaling, tumor angiogenesis and myeloid cell migration in vivo.
Specifications
Specifications
| Chemical Name or Material | 5-Chloro-N2-[(1S)-1-(5-fluoro-2-pyrimidinyl)ethyl]-N4-(5-methyl-1H-pyrazol-3-yl)-2,4-pyrimidine-2,4-diamine |
| CAS | 935666-88-9 |
| Quantity | 10 mg |
| Target | JAK Kinase Inhibitors |
| Molecular Formula | C14H14ClFN8 |
| Formula Weight | 348.77 |
| Purity | >98% |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction