Learn More
Azathioprine-13C4, TRC
Specifications
Specifications
| Chemical Name or Material | Azathioprine-13C3 |
| CAS | 1346600-71-2 |
| Molecular Formula | 13C4 C5 H7 N7 O2 S |
| Quantity | 0.5 mg |
| Synonym | Azathioprine 13C4 |
| InChI Formula | InChI=1S/C9H7N7O2S/c1-15-4-14-7(16(17)18)9(15)19-8-5-6(11-2-10-5)12-3-13-8/h2-4H,1H3,(H,10,11,12,13)/i1+1,4+1,7+1,9+1 |
| SMILES | [13CH3]n1[13cH]n[13c]([13c]1Sc2ncnc3[nH]cnc23)[N+](=O)[O-] |
| IUPAC Name | 6-(3-(1^{13}C)methyl-5-nitro(2,4,5-^{13}C_{3})imidazol-4-yl)sulfanyl-9H-purine |
| Molecular Weight (g/mol) | 281.23 |
| Formula Weight | 281.0516 |