missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ AZ' '9567
Description
AZ'9567 is a potent MAT2a (methionine adenosyltransferase 2a) inhibitor (IC50 = 1.3 nM). AZ'9567 shows strong antiproliferative effect on methylthioadenosine phosphorylase (MTAP) null lymphoma, lung, and pancreatic cancer cell lines and in vivo in MTAP KO HCT116 xenograft mouse model (20 mg/kg twice a day). AZ'9567 is orally bioavailable.
Specifications
Specifications
| Chemical Name or Material | 3-Cyclopropyl-4-[4-(difluoromethoxy)phenyl]-1,4-dihydro-6-(2-methyl-2H-indazol-5-yl)-5H-pyrazolo[4,3-b]pyridin-5-one |
| CAS | 3056570-19-2 |
| Quantity | 5 mg |
| Product Line | Tocris |
| Target | Other Transferase Inhibitors |
| Molecular Formula | C24H19F2N5O2 |
| Purity | 98% |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?