missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ AZ 3137
Description
AZ 3137 is a potent and selective androgen receptor (AR) Degrader (PROTAC®)(DC50 = 22 nM in LNCaP cells). AZ 3137 can degrade L702H mutant AR (DC50 = 158 nM). AZ 3137 inhibits the proliferation of LNCaPs cells (GI50 = 74 nM). AZ 3137 inhibits AR signaling in vitro and tumor growth in vivo in a mouse prostate cancer xenograft model. AZ 3137 is orally bioavailable.
Specifications
Specifications
| Chemical Name or Material | 4-[4-[4-[4-[[4-[2-(2,6-Dioxo-3-piperidinyl)-2,3-dihydro-7-methoxy-1-oxo-1H-isoindol-5-yl]-1-piperazinyl]methyl]-1-piperidinyl]phenyl]-1-piperidinyl]-3-fluoro-2-(trifluoromethyl)benzonitrile |
| CAS | 2960179-55-7 |
| Quantity | 5 mg |
| Product Line | Tocris |
| Target | Active Degraders |
| Molecular Formula | C43H47F4N7O4 |
| Purity | 98% |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?