Learn More
R&D Systems™ AZ 14145845
Description
AZ 14145845 is a potent, and highly selective dual Mer/Axl kinase inhibitor (pIC50 = 7 nM and 7.8 nM in pAxl and pMer, respectively). In vivo, AZ 14145845 shows dose-dependent reduction of tumor growth in the Mer and Axl kinase Ba/F3 tumor xenograft model. AZ 14145845 combined with anti-PD1 antibodies and ionizing radiation increases survival in mice with MC38 tumors in vivo. AZ 14145845 is orally bioavailable.
Specifications
Specifications
| Chemical Name or Material | 3-[4-[(Dimethylamino)methyl]phenyl]-N-methyl-N-[5-[3-methyl-5-(1,3,5-trimethyl-1H-pyrazol-4-yl)-2-pyridinyl]-1,3,4-oxadiazol-2-yl]imidazo[1,2-a]pyridine-6-methanamine |
| CAS | 2830555-70-7 |
| Quantity | 50 mg |
| Product Line | Tocris |
| Target | Additional RTK Inhibitors |
| Molecular Formula | C32H35N9O |
| Purity | 0.97 |