Learn More
Axygen™ AxyMats™ Sealing Mats for 500uL 96 Well Plates
Shop All Axygen Scientific Products
Click to view available options
Pre-slit:
No
Sterility:
Non-sterile
Sterile
Unit Size:
Pack of 50
Description
Axygen AxyMat sealing mats can be used in a wide range of temperatures for PCR and storage applications. AxyMats minimize evaporation and are made of research-grade silicone which can be autoclaved. They can be reused following a bleach wash and ethanol rinse protocol.
Specifications
Specifications
| Color | Natural |
| Material | Silicone |
| Sterility | Sterile |
| Autoclavable | Autoclavable |
| No. of Wells | 96 |
| No. per Case | 5 |
| No. per Pack | 10 |
| Pre-slit | No |
| Product Type | Sealing Mat |
| Plate Compatibility | 96-well Deep Well Plate, 0.5 mL |