Learn More
Axygen™ Automation Micropipette Tips
Product Code. 12716679 Shop All Axygen Scientific Products
Click to view available options
Sterility:
Non-sterile
Sterile
Tip Style:
Automation, Maxymum Recovery, Round
Automation, Round
Unit Size:
Pack of 4800
Product Code. 12716679
Brand: Axygen™ ZT200LRS
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
Axygen™ Automation Micropipette Tips are constructed of polypropylene, and feature a variety of available options to suit your needs.
  • For Tecan™/Freedom EVO&trade, Te-Mo™, Aquarius, Zymark™/Caliper, RapidPlate™, and SciClone™ Systems
  • For 96-well, Non-ANSI and ANSI/SLAS (384) format, number 5 polypropylene racks
  • Available with or without aerosol-resistant filters
  • Available sterile/non-sterile
  • Available in standard or Maxymum Recovery™ style
Order Info
96 Tips/Rack, 10 Racks/Pack, 5 Packs/Case
TRUSTED_SUSTAINABILITY
Specifications
Volume (Metric) 200 μL
Filtered Non-filtered
Sterility Sterile
Color Clear
Rack Color Blue
Autoclavable Autoclavable
Clean Claims RNase-free, DNase-free, Nonpyrogenic
Format Rack
Tip Style Automation, Maxymum Recovery, Round
Packaging 96 tips/rack, 10 racks/pack, 5 packs/case
Quantity 50 Racks/Cs.
Product Type Automation Micropipette Tip
For Use With (Equipment) Tecan/Freedom EVO, Te-Mo, Aquarius, Zymark/Caliper, RapidPlate, and SciClone Systems