Learn More
Axygen™ Black Hanging Format Automation Pipet Tips for Hamilton Microlab™ Star Line and Nimbus™
BIOPHARMAGREENER_CHOICE
Product Code. 12822283 Shop All Axygen Scientific Products
Change view
Click to view available options
Volume (Metric):
10 μL
50 μL
300 μL
1000 μL
Sterility:
Non-sterile
Sterile
Filtered:
Filtered
Non-filtered
Format:
Hanging Tip Rack
Stack Rack
Unit Size:
Pack of 11520
Pack of 1536
Pack of 2304
15 product options available for selection
Product selection table with 15 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) Sterility Filtered Format unitSize
12822283 10 μL Non-sterile Non-filtered Hanging Tip Rack Pack of 2304
12842283 10 μL Sterile Non-filtered Hanging Tip Rack Pack of 2304
12852293 10 μL Non-sterile Non-filtered Stack Rack Pack of 11520
12832283 10 μL Sterile Filtered Hanging Tip Rack Pack of 2304
12862283 50 μL Non-sterile Non-filtered Hanging Tip Rack Pack of 2304
12852283 50 μL Sterile Non-filtered Hanging Tip Rack Pack of 2304
12892283 50 μL Sterile Filtered Hanging Tip Rack Pack of 2304
12892293 50 μL Non-sterile Non-filtered Stack Rack Pack of 11520
12882283 300 μL Non-sterile Non-filtered Hanging Tip Rack Pack of 2304
12872283 300 μL Sterile Non-filtered Hanging Tip Rack Pack of 2304
12832303 300 μL Non-sterile Non-filtered Stack Rack Pack of 11520
12802293 300 μL Sterile Filtered Hanging Tip Rack Pack of 2304
12832293 1000 μL Sterile Filtered Hanging Tip Rack Pack of 1536
12822293 1000 μL Sterile Non-filtered Hanging Tip Rack Pack of 1536
12812293 1000 μL Non-sterile Non-filtered Hanging Tip Rack Pack of 1536
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 15 options available.
15 options
Product Code. 12822283 Supplier Axygen™ Supplier No. HT10CBKHTR
Estimated Shipment: 17-12-2026
Add to basket
Add to basket
Compatible with Hamilton MICROLAB™ STAR and Nimbus workstations
Axygen offers a comprehensive line of tips for Hamilton™ Microlab™ automation systems.
The tips are:
  • Manufactured to stringent specifications under strict process controls and assembled via automation to ensure consistent performance and quality
  • RNase-/DNase-free and non-pyrogenic
  • Available in non-sterile, sterile, non-filtered and filtered
  • Conductive black polypropylene tips are available in 96-well hanging tip rack and nested tip rack and individual deck-seating rack format
  • Designed for Hamilton™ MICROLAB™ STAR, STARlet, STARplus and Nimbus™ workstations.
TRUSTED_SUSTAINABILITY
Specifications
Volume (Metric) 10 μL
Filtered Non-filtered
Sterility Non-sterile
Color Black
Autoclavable Not Autoclavable
Clean Claims DNase, RNase and Pyrogen free
For Use With (Application) The conductive, liquid level sensing tips are available in four sizes and specially designed for use with Hamilton™ liquid handling platforms, they enable precision pipetting across a wide range of applications
Format Hanging Tip Rack
Tip Style Automation, Round
Packaging 1 Rack/Pk, 24 Pks/Cs.
For Use With (Equipment) Hamilton MICROLAB STAR, Hamilton MICROLAB STARlet, Hamilton MICROLAB STARplus, Hamilton MICROLAB Nimbus