Learn More
Axygen™ Polypropylene Single Well Reagent Reservoir
Shop All Axygen Scientific Products
Click to view available options
Certifications/Compliance:
Certified RNase-/DNase-free
Unit Size:
Pack of 25
Description
Corning offers a complete line of Axygen single-well and multiple-well reagent reservoirs which are available in multiple formats for automated and manual use. All reservoirs are designed to comply with standard ANSI/SLAS footprint dimensions for microplates and are compatible with most automation workstations. Reagent reservoirs are manufactured with polypropylene for excellent chemical resistance.
- Durable and cost effective
- Various formats for multiple applications
Specifications
Specifications
| Certifications/Compliance | Certified RNase-/DNase-free |
| Color | Clear |
| Material | Polypropylene |
| Sterility | Sterile |
| Type | Reagent Reservoir |
| No. per Case | 25 |
| No. per Pack | 25 |
| Channels | 8 |
| Bottom Shape | Flat |