Learn More
Axygen™ PlateMax™ Semi-Automated Plate Sealer Accessory, Heat Sealing Films
Product Code. 12779618 Shop All Axygen Scientific Products
Change view
Click to view available options
Product Type:
Heat Sealing Film for Compound Storage
Heat Sealing Film for qPCR
Peelable Heat Sealing Film
Peelable Heat Sealing Film for qPCR
Pierceable Aluminum Heat Sealing Film
Unit Size:
Pack of 500
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Product Type Unit Size
12779618 Peelable Heat Sealing Film for qPCR Pack of 500
12769618 Heat Sealing Film for qPCR Pack of 500
12647765 Heat Sealing Film for Compound Storage Pack of 500
12627765 Peelable Heat Sealing Film Pack of 500
12637765 Pierceable Aluminum Heat Sealing Film Pack of 500
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Product Code. 12779618 Supplier Axygen™ Supplier No. HS150QPCR
  • ONLINE EXCLUSIVE PRICE
    620.00€
    418.28€ / Pack of 500
    Save 201.72€
    32% Off
View all promotions
Estimated Shipment: 09-04-2027
Designed for applications ranging from PCR and Real-Time PCR to ELISA and cell culture
Axygen PlateMax microplate heat sealing films are designed for applications ranging from PCR and Real-time PCR to storage. They can be used in a wide range of temperatures making them suitable for nearly any application.
TRUSTED_SUSTAINABILITY
Spezifikation
For Use With (Application) Plate Sealing
For Use With (Equipment) PlateMax Plate Sealer
Material Polyester
No. per Case 500/Cs.
No. per Pack 100/Pk.
Product Type Peelable Heat Sealing Film for qPCR