Learn More
Axygen™ Low Profile 8-Strip PCR Tubes
GREENER_CHOICE
Product Code. 15153727 Shop All Axygen Scientific Products
Change view
Click to view available options
Color:
Clear
White
Product Type:
Thin Wall 8-Strip PCR Tube
Thin Wall 8-Strip PCR Tube with Real Time Strip Caps
Unit Size:
Pack of 125
Pack of 1250
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color Product Type unitSize
15153727 Clear Thin Wall 8-Strip PCR Tube Pack of 125
15133707 Clear Thin Wall 8-Strip PCR Tube with Real Time Strip Caps Pack of 125
13419818 Clear Thin Wall 8-Strip PCR Tube with Real Time Strip Caps Pack of 1250
13429818 White Thin Wall 8-Strip PCR Tube with Real Time Strip Caps Pack of 1250
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 15153727 Supplier Axygen™ Supplier No. PCR0108LPC
Add to basket
Add to basket
Axygen™ brand low profile 8-strip PCR tubes are ideal for low sample volume reactions.
Axygen™ Low Profile 8-Strip PCR tubes are ideal for low sample volume reactions. Using highly polished and precise molds, our tubes ensure precise thermal transfer and a leak-proof seal with a choice of strip caps. The low profile design allows reduced reaction time and evaporation due to less dead air volume. Tubes are available in white and clear with or without caps. Tubes are DNase-/RNase-free and nonpyrogenic.
  • Low profile design results in faster and more efficient PCR reactions
  • Ideal for 0.1 mL thermal cyclers
  • Available in white or clear
  • DNase-/RNase-free and Nonpyrogenic
  • Recommended volume range = 5 μL to 125 μL
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance RNase, DNase and non-pyrogenic free
Color Clear
Volume (Metric) 0.1 mL
Quantity 125/Pk.
For Use With (Equipment) 0.1 mL thermal cycler for low sample volume reactions
Shape Conical
Sterility Non-sterile
Material Polypropylene
Format Strip
Product Type Thin Wall 8-Strip PCR Tube