Learn More
Axygen™ Gel Documentation System Accessory, Orange Glass
Product Code. 16603014 Shop All Axygen Scientific Products
Change view
Click to view available options
For Use With (Equipment):
Gel Documentation System (GD-1000)
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) Unit Size
16603014 Gel Documentation System (GD-1000) Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 16603014 Supplier Axygen™ Supplier No. GDOG
  • ONLINE EXCLUSIVE PRICE
    163.00€
    161.18€ / Pack of 1
    Save 1.82€
    1% Off
Estimated Shipment: 11-03-2027
For use with Gel Documentation System (GD-1000)
Ideal for reducing background when cutting out bands with blue light
TRUSTED_SUSTAINABILITY
Specifications
Product Type Orange Glass
For Use With (Equipment) Gel Documentation System (GD-1000)