Learn More
Axygen™ Conical-Bottom Screw-Cap Tubes, Attached Clear Looped Caps
Shop All Axygen Scientific Products
Click to view available options
Color:
Clear
Unit Size:
Pack of 500
Description
- Writing area for sample identification
- Caps close with one hand
Specifications
Specifications
| Cap Style | Screw Cap |
| Autoclavable | Not Autoclavable |
| Capacity (Metric) | 1.5 mL |
| Color | Clear |
| Material | Polypropylene |
| For Use With (Application) | For sample identification |
| Includes | Attached clear looped cap |
| Sterility | Non-sterile |
| Bottom Shape | Conical |
| Max. RCF | 20,000 x g |