Learn More
Axygen™ Caps for PCR Tubes
Product Code. 12881510 Shop All Axygen Scientific Products
Click to view available options
Color:
Assorted
Clear
Green
MaxyClear
MaxyClear, w/writing surface
Orange
Red
Violet
Yellow
Closure Type:
Domed Cap
Flat Cap
Product Type:
8-Strip Domed PCR Tube Cap
8-Strip Flat PCR Tube Cap
8-Strip Flat PCR Tube Cap with Writing Surface
PCR Caps
Unit Size:
Pack of 100
Pack of 125
Pack of 1250
Product Code. 12881510
Brand: Axygen™ PCR02CPO
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
8-Strip domed or flat strip caps for Axygen PCR tubes
Axygen polypropylene, thin wall PCR tubes with and without caps are designed to fit standard 96-well blocks and are available in an assortment of colors. The consistent and ultra-thin wall allows precise thermal transfer. An even insertion cap ensures a tight fit to reduce evaporation.
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance RNase, DNase, pyrogen-free
Closure Color Orange
Color Orange
Volume (Metric) 0.2 mL
Quantity 125/Pk.
Closure Material Polypropylene
For Use With (Equipment) 8-strips PCR tubes and 96-well PCR microplates
Sterility Non-sterile
Material Polypropylene
Closure Type Domed Cap
Format Strip
Product Type 8-Strip Domed PCR Tube Cap