Learn More
Axygen™ Caps for PCR Tubes
Shop All Axygen Scientific Products
Click to view available options
Color:
Assorted
Clear
Green
MaxyClear
MaxyClear, w/writing surface
Orange
Red
Violet
Yellow
Closure Type:
Domed Cap
Flat Cap
Product Type:
8-Strip Domed PCR Tube Cap
8-Strip Flat PCR Tube Cap
8-Strip Flat PCR Tube Cap with Writing Surface
PCR Caps
Unit Size:
Pack of 100
Pack of 125
Pack of 1250
Beschreibung
Axygen polypropylene, thin wall PCR tubes with and without caps are designed to fit standard 96-well blocks and are available in an assortment of colors. The consistent and ultra-thin wall allows precise thermal transfer. An even insertion cap ensures a tight fit to reduce evaporation.
Spezifikation
Spezifikation
| Certifications/Compliance | RNase, DNase, pyrogen-free |
| Closure Color | Assorted |
| Color | Assorted |
| Volume (Metric) | 0.2 mL |
| Quantity | 125/Pk. |
| Closure Material | Polypropylene |
| For Use With (Equipment) | 8-strips PCR tubes and 96-well PCR microplates |
| Sterility | Non-sterile |
| Material | Polypropylene |
| Closure Type | Domed Cap |
| Mehr anzeigen |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur