Learn More
Axygen™ AxyMats™ 96 Round Well Compression Mat for PCR Microplates
Shop All Axygen Scientific ProductsDescription
Axygen™ Compression AxyMatsTM are ideally suited to help reduce evaporation and ensure a proper seal when used in conjunction with Axygen PCR Microplates and PlateMaxTM sealing films. Made of research-grade silicone, they can be autoclaved, and following a bleach wash and ethanol rinse protocol, they can also be reused.
For use with 96/384 well and PCR microplates
Specifications
Specifications
| Color | Natural |
| Material | Silicone |
| Sterility | Sterile |
| Autoclavable | Autoclavable |
| No. of Wells | 96 |
| No. per Case | 50/Cs. |
| No. per Pack | 10/Pk. |
| Product Type | Compression Mat |
| Well Shape | Round |
| Plate Compatibility | 96/384-well PCR Microplate |