Learn More
Axygen™ Automation Tips for STARLine, Clear, CO-RE Style Liquid Level Sensing Tip, Sterile
Shop All Axygen Scientific Products
Click to view available options
Volume (Metric):
10 μL
50 μL
For Use With (Equipment):
Hamilton MICROLAB STAR, Hamilton MICROLAB STARlet, Hamilton MICROLAB STARplus
Hamilton MICROLAB STAR, Hamilton MICROLAB STARlet, Hamilton MICROLAB STARplus, Hamilton MICROLAB Nimbus
Unit Size:
Pack of 11520
Description
Axygen brand automation tips are engineered to be compatible with original equipment manufacturers' tips, ensuring complete compatibility and a consistent fit. One of the widest product offerings combined with our engineering expertise stringent QA/QC enables you to get optimum performance no matter which automated liquid handler you are working with.
- Compatible with Hamilton™ Microlab™ STAR, STARlet, STARplus and Nimbus™
- Conductive black polypropylene
- Certified DNase-/RNase-free, nonpyrogenic
- With or without aerosol-resistant filters
- Nonsterile or sterile
Specifications
Specifications
| Volume (Metric) | 50 μL |
| Filtered | Non-filtered |
| Sterility | Sterile |
| Color | Black |
| Clean Claims | DNase, RNase Free |
| Format | Refill |
| Tip Style | Round |
| For Use With (Equipment) | Hamilton MICROLAB STAR, Hamilton MICROLAB STARlet, Hamilton MICROLAB STARplus |
| Material | Polypropylene |