Learn More
Axygen™ 96-well PCR Microplates
GREENER_CHOICE
Product Code. 15368301 Shop All Axygen Scientific Products
Click to view available options
Skirt Style:
Elevated, Semi
Full
Half
Sterility:
Non-sterile
Barcode:
Includes Barcode
Unit Size:
Pack of 50
Product Code. 15368301
Brand: Axygen™ PCR96M2HSCPBC
Constructed with ultra-thin and consistent wall thickness to allow precise thermal transfer for optimum results. Axygen™ 96-well PCR Microplates are the ideal option for use in PCR thermal transfer and Real-Time PCR applications.
Axygen™ 96 well elevated skirt bar coded PCR microplate is designed to fit ABI™ Real Time PCR thermal cyclers and sequencers. Ultra-thin and consistent wall thickness allows precise thermal transfer for optimum results. Can be sealed with 8/12 strip caps, Axymat or sealing film. Bar code on the long side of the plate between well H1-H12.
  • Certified RNase-/DNase-free
  • Nonpyrogenic
  • Nonsterile
  • Manufactured with 99.9% pure virgin polypropylene
  • 96 Well Semi-Skirted PCR Plate
  • Single Notch Coner
  • Application/Compatibility: Standard thermal cycling, ABI Real time PCR 7300, 7500, 7900, qPCR and ABI Sequencing
  • TRUSTED_SUSTAINABILITY
    Specifications
    Barcode Includes Barcode
    Certifications/Compliance RNase-, DNase-free
    Format Standard
    Array 8 x 12
    Color Clear
    For Use With (Equipment) ABI PCR 2700, 2720, 9700, 9800, 7300, 7500, 7900 and ABI sequencing units
    Skirt Style Half
    Material Polypropylene
    No. of Wells 96
    Sterility Non-sterile
    Well Shape Round
    For Use With (Application) PCR thermal transfer, RT-PCR applications