missing translation for 'onlineSavingsMsg'
Learn More

Axiom™ GeneTitan™ Consumables Kit

Product Code. 30826223 Shop All Applied Biosystems Products
Change view
Click to view available options
Quantity:
1 plate
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30826223 1 plate
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30826223 Supplier ABI Applied Biosystems Supplier No. 901606
 Restricted Item
In Stock
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Axiom™ GeneTitan™ Consumables Kit includes all GeneTitan instrument consumables required to process one Axiom 96 array plate.

TRUSTED_SUSTAINABILITY

Specifications

Type Consumables Kit
Format 96-array Plate
Content And Storage Store all at room temperature
Contains:
5 X 96 layout stain trays
1 X 96 layout Hybridization tray
1 X 96 layout Scan tray
6 X 96 layout Scan and Stain tray covers
Quantity 1 plate
Reagent Type Genetitan consumables

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.