Learn More
Zeiss™ Axiolab 5 Upright Microscope for Transmitted Light Brightfield and Phase Contrast
Product Code. 17302820
Change view
Click to view available options
For Use With (Application):
Transmitted Light Brightfield and Phase Contrast
Unit Size:
1 set
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Application) Unit Size
17302820 Transmitted Light Brightfield and Phase Contrast 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 17302820 Supplier Zeiss™ Supplier No. 4909800002000
  • ONLINE EXCLUSIVE PRICE
    9195.50€
    8096.00€ / 1 sets
    Save 1099.50€
    11% Off
 Restricted Item
Estimated Shipment: 06-11-2026
Add to basket
Add to basket
Compact and ergonomic upright microscope for routine lab work. This system is configured for transmitted brightfield and phase contrast microscopy with a photo binocular tube. Combine this microscope with the Axiocam 208 to take full advantage of SMART microscopy for easy digital documentation.
  • Axiolab 5 is made for the routine microscopy work that goes on every day in your lab. Its compact and ergonomic design saves space and makes for easy handling.
  • Combine it with Axiocam 208 color for SMART microscopy. Just focus your sample and press a single button for crisp images in true color. The digital image will look like you see it through the eyepieces, with all the details and subtle color differences clearly visible. Plus, Axiolab 5 automatically adds the correct scaling information to your images. 
  • Microscope body is powder coated durable metal, heat treated and resistant to standard reagents used for staining, organic solvents used for cleaning of lenses and disinfectants.
  • All optical parts including objectives, eye pieces, lenses, prisms, have anti-reflective and anti-fungal coating.
  • Possible contrast methods are brightfield, darkfield, phase contrast, and simple transmitted light polarization. Upgrades from brightfield to other transmitted light contrasting techniques are available. With Axiolab 5 FL fluorescence is available in addition.
  • The microscope comes with a manual revolving nosepiece with 5 encoded positions, suitable for reading objective position for light intensity manager functionality and for scaling information
Specifications
For Use With (Application) Transmitted Light Brightfield and Phase Contrast