Learn More
Thermo Scientific™ Avermectin, ACROS Organics™
Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
Specifications
| Chemical Name or Material | Avermectin |
| CAS | 65195-55-3 |
| Quantity | 250 mg |
| Molecular Formula | C48 H72 O14 |
| Formula Weight | 873.09 |
| Physical Form | Crystalline Powder |
| Color | White to Light-yellow |