missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals AutoScan Pro 4.0

Product Code. 30694930 Shop All MP Biomedical Products
Click to view available options
Type:
Software
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 30694930

Brand: MP Biomedicals EMC021

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

AutoScan Pro is a software which enables an automated analysis and interpretation of Western Blot and Line Immunoblot test results.

  • AutoScan Pro 4.0 reads the digital image of the strips generated by a validated scanner.
  • Using the pre-programmed interpretation criteria recommended by MP Biomedicals, AutoScan Pro 4.0 evaluates the test strip results and provides an accurate interpretation to the user.
  • AutoScan Pro quantifies the band intensity and interprets the results, thus preventing any potential human error.
  • Fast and Efficient: analysis and interpretations of many strips in a few minutes.
  • Highly accurate: quantification of band intensity to interprete the results.
  • Reliable: results analysis and interpretation are highly reproducible.
  • Significantly low workload.

Specifications

For Use With (Equipment) AutoBlot System 36, Autoblot System 48
Type Software
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.