Learn More
- Home
- Products
- Chromatography Syringes
- Autosampler Syringes
- Trajan™ Autosampler Syringes with Removable Needle for CTC Analytics CombiPAL and GC PAL
Trajan™ Autosampler Syringes with Removable Needle for CTC Analytics CombiPAL and GC PAL
Code produit 13199205
Modifier l'affichage
Click to view available options
Needle Gauge:
23 G
26 G
Diameter (English) Outer Needle:
0.01 in.
0.018 in.
0.02 in.
Length (English) Needle:
1.9 in.
2.2 in.
3.3 in.
Model:
10F-RTC/RSH-5.7/0.47C
10F-RTC/RSH-5.7/0.63C
10F-RTC/RSH-8.5/0.47C
10F-RTC/RSH-8.5/0.63C
10F-RTC/RSH-GT-5.7/0.47C
10F-RTC/RSH-GT-5.7/0.63C
10F-RTC/RSH-GT-8.5/0.47C
10F-RTC/RSH-GT-8.5/0.63C
25F-RTC/RSH-GT-5.7/0.47C
25F-RTC/RSH-GT-5.7/0.63C
25F-RTC/RSH-GT-8.5/0.63C
25FRTC/RSH-GT-8.5/0.47C
25R-C/T-GT-0.47
50F-RTC/RSH-GT-5.7/0.47C
50F-RTC/RSH-GT-5.7/0.63C
50F-RTC/RSH-GT-8.5/0.47C
50F-RTC/RSH-GT-8.5/0.63C
5F-RTC/RSH-5.7/0.47C
5F-RTC/RSH-5.7/0.63C
5F-RTC/RSH-8.5/0.63C
Conditionnement:
Each
20 product options available for selection
| Code produit | Needle Gauge | Diameter (English) Outer Needle | Length (English) Needle | Model | unitSize |
|---|---|---|---|---|---|
| 13199205 | 23 G | 0.02 in. | 2.2 in. | 50F-RTC/RSH-GT-5.7/0.63C | Each |
| 13189015 | 26 G | 0.01 in. | 2.2 in. | 10F-RTC/RSH-5.7/0.47C | Each |
| 13186105 | 23 G | 0.02 in. | 2.2 in. | 5F-RTC/RSH-5.7/0.63C | Each |
| 13199195 | 26 G | 0.01 in. | 2.2 in. | 10F-RTC/RSH-GT-5.7/0.47C | Each |
| 13179015 | 23 G | 0.02 in. | 3.3 in. | 10F-RTC/RSH-8.5/0.63C | Each |
| 13109205 | 26 G | 0.01 in. | 3.3 in. | 10F-RTC/RSH-GT-8.5/0.47C | Each |
| 13109215 | 23 G | 0.02 in. | 3.3 in. | 50F-RTC/RSH-GT-8.5/0.63C | Each |
| 13106115 | 26 G | 0.01 in. | 2.2 in. | 5F-RTC/RSH-5.7/0.47C | Each |
| 13169015 | 23 G | 0.02 in. | 2.2 in. | 10F-RTC/RSH-5.7/0.63C | Each |
| 13196105 | 23 G | 0.02 in. | 3.3 in. | 5F-RTC/RSH-8.5/0.63C | Each |
| 13129215 | 26 G | 0.01 in. | 3.3 in. | 50F-RTC/RSH-GT-8.5/0.47C | Each |
| 13179195 | 23 G | 0.02 in. | 2.2 in. | 10F-RTC/RSH-GT-5.7/0.63C | Each |
| 13189205 | 26 G | 0.01 in. | 3.3 in. | 25FRTC/RSH-GT-8.5/0.47C | Each |
| 13179205 | 26 G | 0.01 in. | 2.2 in. | 25F-RTC/RSH-GT-5.7/0.47C | Each |
| 13119215 | 26 G | 0.01 in. | 2.2 in. | 50F-RTC/RSH-GT-5.7/0.47C | Each |
| 13189195 | 23 G | 0.02 in. | 3.3 in. | 10F-RTC/RSH-GT-8.5/0.63C | Each |
| 13159205 | 23 G | 0.02 in. | 2.2 in. | 25F-RTC/RSH-GT-5.7/0.63C | Each |
| 13169205 | 23 G | 0.02 in. | 3.3 in. | 25F-RTC/RSH-GT-8.5/0.63C | Each |
| 13199015 | 26 G | 0.01 in. | 3.3 in. | 10F-RTC/RSH-8.5/0.47C | Each |
| 12977506 | 26 G | 0.018 in. | 1.9 in. | 25R-C/T-GT-0.47 | Each |
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 20 options available.
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
- Borosilicate glass barrel for strength and inertness.
- Stainless steel needles for inertness, strength and corrosion resistance.
- Backing behind the scale aids quick and accurate reading of the syringe scale.
- Metal back flange provides stability and resists fracture.
- Metal needle cap on syringe barrel protects and improves syringe lifetime.
- Syringes are individually tested for quality ensuring reliability and conformance.
- Unique packaging design eliminates possible contamination in the syringe.
Spécification
Spécification
| Needle Gauge | 23 G |
| Diameter (English) Outer Needle | 0.02 in. |
| Diameter (Metric) Outer Needle | 0.63 mm |
| For Use With (Application) | Gas chromatography |
| Length (English) Needle | 2.2 in. |
| Product Type | Autosampler Syringe |
| Model | 50F-RTC/RSH-GT-5.7/0.63C |
| Quantity | Ea. |
| Volume (Metric) | 50 μL |
| Needle Point Style | Cone |
| For Use With (Equipment) | Thermo RSH Autosampler |
| Connection Type | Fixed |
| Afficher plus Afficher moins |
Nom du produit
Sélectionnez un problème
En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.
Vous avez repéré une opportunité d'amélioration ?