Learn More
Merck Autorad Orientation Marker
Shop All Merck Products
Click to view available options
Quantity:
5 Pk.
Unit Size:
Pack of 5
Description
- For accurate orientation of an autoradiograph to its membrane
- Emits low-level luminescence rather than radioactivity
- Safe, non-hazardous alternative to radioactive ink
Specifications
Specifications
| Product Type | Orientation Marker |
| For Use With (Application) | Used in Western Blotting, dot/slot Blotting, For Use on Immunoblots |
| Quantity | 5 Pk. |