Learn More
Applied Biosystems™ AutoMate Express™ Nucleic Acid Extraction System
Product Code. 30055377 Shop All Applied Biosystems Products
Change view
Click to view available options
Quantity:
1 instrument
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30055377 1 instrument Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30055377 Supplier Applied Biosystems™ Supplier No. 4467754
  • ONLINE EXCLUSIVE PRICE
    53340.00€
    51600.00€ / Pack of 1
    Save 1740.00€
    3% Off
 Restricted Item
Estimated Shipment: 11-12-2026
The AutoMate Express Nucleic Acid Extraction System is an easy-to-use benchtop instrument that enables hands-free automation of the nucleic acid extraction process for contaminant and impurity testing.
The AutoMate Express Nucleic Acid Extraction System is an easy-to-use benchtop instrument that enables hands-free automation of the nucleic acid extraction process for contaminant and impurity testing. The instrument uses the established PrepSEQ nucleic acid extraction chemistry for quantitative recovery of DNA and RNA from diverse samples types, including cell culture, raw material (i.e., serum), and in-process downstream purification samples such as column purification or bulk drug substance (BDS) samples.
Features of the AutoMate Express Nucleic Acid Extraction System include:
  • Time-saving, cost-effective DNA extraction for mycoplasma, MMV, and vesivirus
  • Reproducible recovery of DNA from complex samples
  • Out-of-the-box automation with prefilled PrepSEQ Express cartridges and ready-to-run protocols
  • Closed system minimizes the risk of contamination
  • Ideal for in-process testing of contaminants
Three-in-one extraction of cell culture contaminants
The AutoMate Express system can be used to perform nucleic acid extraction of the common contaminants in mammalian cell culture manufacturing, i.e., mycoplasmas, MMV, and vesiviruses (Figure 1). Flexible protocols are available to isolate DNA and RNA from a single organism, or a 3-in-1 surveillance panel protocol can simultaneously extract mycoplasma, MMV, and vesivirus nucleic acid from a single sample.
Closed system with walk-away automation
The AutoMate Express system automatically performs all steps for efficient nucleic acid extraction from contaminants in a closed environment. The system uses PrepSEQ Express kits that contain sealed cartridges with extraction reagents, and plastics, including tips and sample, lysis, and elution tubes. The workflow has been designed for a completely hands-free operation enabling reproducible extraction efficiencies and reduced time-to-results.
TRUSTED_SUSTAINABILITY
Specifications
Product Type Nucleic Acid Extraction System
For Use With (Application) Quantitative recovery of DNA and RNA
Product Line AutoMate Express
Quantity 1 instrument
Samples per Run Up to 13 samples