Learn More
JP Selecta Autoclave Auto Tester "ST DRY PV" -B 23L
Brand:  JP Selecta 4001735
Autoclave Auto Tester "ST DRY PV" -B 23L
Product Code. 15831587
  • ONLINE EXCLUSIVE PRICE
    5405.00€
    3980.00€ / Each
    Save 1425.00€
    26% Off
Only null left
Estimated Shipment: 10-02-2026