Learn More
Cole-Parmer™ Parker Hannifin™ PFA Union Connector Compression Fitting
Product Code. 11702029
Click to view available options
For Use With:
1/4 in. tubing O.D.
1/8 in. tubing O.D.
3/8 in. tubing O.D.
4mm tubing O.D.
6mm tubing O.D.
Shape:
Elbow
Straight
T-shape
Unit Size:
Each
Product Code. 11702029
Brand: Cole-Parmer™ 3132021
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
Compression fittings are an excellent choice to connect rigid tubing systems, able to withstand higher pressures than barbed fittings.
  • Available in straight, "T", and elbow shapes for unmatched versatility
  • Provided with a nut that compresses rigid tubing against the fitting body for high-pressure applications
  • Excellent for use with tubing manufactured from fluoropolymers, other rigid plastics and metals
  • Assemble the fitting to the tubing by sliding the tubing through a sealing ferrule and using the nut to compress down to form a secure seal
  • Sterilization through autoclaving or ethylene oxide (ETO)
Specifications
Autoclavable Autoclavable
Material PFA
For Use With 1/8 in. tubing O.D.
Packaging Quantity 1 Each
Sterilization Method Autoclave or Ethylene Oxide
Shape Straight
Product Type Compression Fitting