Learn More
Abeba™ Autoclavable Clogs 39630
Product Code. 15601278
Click to view available options
Size:
36
37
38
39
40
41
42
43
44
45
46
Unit Size:
1 pair
Product Code. 15601278
Brand: Abeba™ 39630/43
Only null left
Add to basket
Add to basket
Antistatic and autoclavable at 121°C. High slip resistance.
  • CE EN ISO 20347:2012 OB A, E SRA
  • Autoclavable, sterilisable special material
  • Orange
  • Slip resistance outsole sole
  • ESD dissipative according to DIN EN 61340
  • Heel strap foldable, adjustable
  • DOUBLE SIZES, please use even size when placing your order (36, 38, 40, 42, 44, 46)
Specifications
Certifications/Compliance CE EN ISO 20347:2012 OB A, E SRA
Color Orange
Size 43
Sole Features Slip-resistant
Style 39630