Learn More
Abeba™ Autoclavable Clogs 39610
Product Code. 15671248
Change view
Click to view available options
Size:
36
37
38
39
40
41
42
43
44
46
Unit Size:
1 pair
10 product options available for selection
Product selection table with 10 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size Unit Size
15671248 42 1 pair
15611248 36 1 pair
15621248 37 1 pair
15631248 38 1 pair
15641248 39 1 pair
15651248 40 1 pair
15661248 41 1 pair
15681248 43 1 pair
15691248 44 1 pair
15611258 46 1 pair
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 10 options available.
10 options
Product Code. 15671248 Supplier Abeba™ Supplier No. 39610/42
  • ONLINE EXCLUSIVE PRICE
    53.05€
    47.91€ / 1 pairs
    Save 5.14€
    9% Off
View all promotions
Estimated Shipment: 02-12-2026
Antistatic and autoclavable at 121°C. High slip resistance.
  • CE EN ISO 20347:2012 OB A, E SRC
  • Autoclavable, sterilisable special material
  • Blue
  • Slip resistance outsole sole
  • ESD dissipative according to DIN EN 61340
  • Heel strap foldable, adjustable
  • DOUBLE SIZES, please use even size when placing your order (36, 38, 40, 42, 44, 46)
Specifications
Certifications/Compliance CE EN ISO 20347:2012 OB A, E SRC
Color Blue
Size 42
Sole Features Slip-resistant
Style 39610