Learn More
Aurothioglucose 80%, TRC
Product Code. 30006973
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30006973 50 mg 50mg
30134911 5 mg 5mg
30137777 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30006973 Supplier Toronto Research Chemicals Supplier No. TRCA79479050MG
 Restricted Item
Estimated Shipment: 14-12-2026
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 12192-57-3
Molecular Formula C6 H11 Au O5 S
Molecular Weight (g/mol) 392.18
Synonym Gold, [1-(thio-κS)-D-glucopyranosato-κO2]-, D-Glucose, 1-thio-, S-gold deriv. (6CI), Gold, (1-thio-D-glucopyranosato)- (8CI), Gold, (1-thio-D-glucopyranosato-O2,S1)-, D-Glucopyranose, 1-thio-, gold complex, [1-(Thio-κS)-D-glucopyranosato-κO2]gold, (D-Glucopyranosylthio)gold, Aureotan, Auromyose, Aurothioglucose, Aurumine, Authron, Brenol, Glucose, 1-thio-, gold salt (1:1), Glysanol B, Gold thioglucose, Gold, (D-glucopyranosylthio)-, Oronol, Solganal, Solganal B
IUPAC Name gold(1+);(3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)oxane-2-thiolate
SMILES OC[C@H]1OC2[S-][Au+][OH][C@@H]2[C@@H](O)[C@@H]1O
Specifications
Quantity 50 mg
InChI Formula InChI=1S/C6H12O5S.Au/c7-1-2-3(8)4(9)5(10)6(12)11-2;/h2-10,12H,1H2;/q;+1/p-1/t2-,3-,4+,5-,6?;/m1./s1
Formula Weight 391.9993
Chemical Name or Material Aurothioglucose