missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Aurothioglucose 70%-13C6, TRC
Specifications
Specifications
| Chemical Name or Material | Aurothioglucose 13C6 |
| Molecular Formula | 13C6H11AuO5S |
| Quantity | 2 mg |
| Synonym | [1-(thio-κS)-D-Glucopyranosato-κO2]-gold-13C6, 1-thio-D-Glucose S-gold Derivative-13C6, (1-thio-D-Glucopyranosato)-Gold-13C6, (1-thio-D-Glucopyranosato-O2,S1)gold-13C6, 1-thio-D-Glucopyranose Gold Complex-13C6, (D-Glucopyranosylthio)gold-13C6, Aureotan-13C6, Auromyose-13C6, Aurothioglucose-13C6, Aurumine-13C6, Authron-13C6, Brenol-13C6, 1-thio-Glucose Gold Salt (1:1)-13C6, Glysanol B-13C6, Gold Thioglucose-13C6, (D-Glucopyranosylthio)gold-13C6, Oronol-13C6, Solganal-13C6, Solganal B-13C6 |
| InChI Formula | InChI=1S/C6H12O5S.Au/c7-1-2-3(8)4(9)5(10)6(12)11-2;/h2-10,12H,1H2;/q;+1/p-1/t2-,3-,4+,5-,6+;/m1./s1/i1+1,2+1,3+1,4+1,5+1,6+1; |
| SMILES | O[13C@@]1([H])[13C@@](O)([H])[13C@](O)([H])[13C@@](S[Au])([H])O[13C@]1([H])[13C]([H])([H])O |
| IUPAC Name | gold(1+);(2S,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxy(113C)methyl)(2,3,4,5,6-13C5)oxane-2-thiolate |
| Molecular Weight (g/mol) | 398.14 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?