Learn More
3M™ Aura™ 9300 Series Particulate Respirators
Product Code. 11757574
Click to view available options
:
Filter Type: FFP3
Product Code. 11757574
Brand: 3M™ 7000088812
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
Providing a unique three-panel design for greater comfort and easy communication. 3M™ Aura™ 9300 Series Particulate Respirators cover a range of applications requiring FFP1, FFP2 and FFP3 protection.
  • Individually packaged to avoid contamination
  • Soft cover web on inner panel ensures greater comfort
  • Lightweight and foldable
  • Evenly tensioned headbands to relieve strap pressure
  • Centrally positioned cool flow valve reduces heat build-up and makes breathing easier and cooler
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance EN149: 2001 A1: 2009 FFP3 NR D
Color Code Red
Design Type 3-panel
Filter Type FFP3
Product Type Dust Mask
With Exhalation Valve Yes
For Use With (Application) General industry, cement dust, pharmaceuticals, pottery and ceramics, career exploitation
Model 9330 Dust Mask
Quantity 20/Pk.
Resistant To Fine dust, water and oil mist
Shape Pliable