Learn More
3M™ Aura™ 9322+ Particulate Respirators
Provides protection against fine dusts and water-based mists.
Brand: 3M™ 7100004175
Additional Details : Weight : 0.80000kg
Description
- Unique three-panel design for greater comfort and easy communication
- Soft cover web on inner panel ensures greater comfort
- Centrally positioned cool flow valve reduces heat build-up and makes breathing easier and cooler
- Evenly tensioned headbands to relieve strap pressure
- Lightweight and foldable
- Individually packed to avoid contamination during storage
- FFP2 level protection
- Particulates Assigned Protection Factor of 10 (NPF 12)