missing translation for 'onlineSavingsMsg'
Learn More

(±)-Atropine-D3, TRC

Product Code. 30335716
Change view
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30335716 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30335716 Supplier Toronto Research Chemicals Supplier No. TRCA7946325MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Atropine D3 (N-methyl D3)
CAS 60365-55-1
Purity Grade Notes HPLC
Molecular Formula C17 2H3 H20 N O3
Quantity 5 mg
Synonym Benzeneacetic acid, α-(hydroxymethyl)-, 8-(methyl-d3)-8-azabicyclo[3.2.1]oct-3-yl ester, endo- (9CI), Benzeneacetic acid, α-(hydroxymethyl)-, 8-(methyl-d3)-8-azabicyclo[3.2.1]oct-3-yl ester, endo-(±)- (ZCI), α-(Hydroxymethyl)benzeneacetic acid endo-8-(methyl-d3)-8-azabicyclo[3.2.1]oct-3-yl ester, Atropine-(N-methyl-D3), Atropine D3 (N-methyl D3), (±)-Atropine-d3 (N-methyl-d3), (±)-Atropine-D3
InChI Formula InChI=1S/C17H23NO3/c1-18-13-7-8-14(18)10-15(9-13)21-17(20)16(11-19)12-5-3-2-4-6-12/h2-6,13-16,19H,7-11H2,1H3/t13-,14+,15+,16-/m0/s1/i1D3
SMILES [2H]C([2H])([2H])N1[C@@H]2CC[C@H]1C[C@H](C2)OC(=O)[C@@H](CO)c3ccccc3
IUPAC Name [rac-(1R,5S)-8-(trideuteriomethyl)-8-azabicyclo[3.2.1]octan-3-yl] rac-(2R)-3-hydroxy-2-phenylpropanoate
Molecular Weight (g/mol) 292.3879
Formula Weight 292.1866
Percent Purity >95
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.