missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ ATF2 (Human) Recombinant Protein
Human ATF2 full-length ORF ( AAH26175, 1 a.a. - 209 a.a.) recombinant protein with GST-tag at N-terminal.
Brand: Abnova™ H00001386-P01.10ug
Additional Details : Weight : 0.00010kg
Description
- Sequence: MKFKLHVNSARQYKDLWNMSDDKPFLCTAPGCGQRFTNEDHLAVHKHKHEMTLKFGPARNDSVIVADQTPTPTRFLKNCEEVGLFNELASPFENEFKKASEDDIKKMPLDLSPLATPIIRSKIEEPSVVETTHQDSPLPHPESTTSDEKEVPLAQTAQPTSAIVRPASLQVPNVLLTSSDSSVIIQQAVPSPTSSTVITQAPSSNRPIV
Specifications
AAH26175 | |
activating transcription factor 2 | |
125% SDS-PAGE Stained with Coomassie Blue | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
ATF2 | |
GST |
1386 | |
Wheat germ expression system | |
10 μg | |
CRE-BP1, CREB2, HB16, MGC111558, TREB7 | |
Wheat Germ (in vitro) | |
50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer |