missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Promed Bioscience Atelocollagen, Porcine Type I (Enzymatic extraction)
Description
- Lyophilized, highly purified Type I atelocollagen with superior functionality and physical/mechanical properties (such as tensile strength and elasticity)
- Proprietary enzyme-based extraction removes telopeptides at the non-helical N and C termini, resulting in a triple helix configuration that exhibits reduced immunogenicity and enhanced biocompatibility
- Suitable for in vitro cell cultures, for cell functional assays, as a chemoattractant agent and for soft 3D cell cultures (e.g. tumor assays)
Specifications
Specifications
| Formulation | Lyophilized Fibrous Powder |
| Purification Method | Dialysis |
| Quantity | 10 mg |
| Content And Storage | -20°C to 8°C |
| Endotoxin Level | <1.0 EU/mL |
| Purity or Quality Grade | >95% (by SDS/Western blotting) |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?