Learn More
ATAGO™ Hand-Held Refractometer, MASTER-3T
Description
- Made of metal
- Equipped with Automatic Temperature Compensation feature Calibration with 60% Sucrose Solution (RE-110060) or Standard liquid LK (RE-99010)
Specifications
Specifications
| Type | Handheld Refractometer |
| Scale Range | 58.0 to 90.0 Brix with ATC |
| Model | MASTER-3T |
| Accuracy | Brix : 0.2% |
| Repeatability | ±0.1% |
| Scale Type | Brix |