Learn More
Thermo Scientific™ Assembled Caps and Septa for 8-425 Standard-Opening Vials
Product Code. 12430138
Click to view available options
Color:
Black
Blue
Natural
Red
White
Yellow
Unit Size:
Pack of 100
Product Code. 12430138
Brand: Thermo Scientific™ C401361A
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
Thermo Scientific™ National Assembled Caps and Septa for 8-425 Standard-Opening Vials are available in a variety of colors and septum materials.
Thermo Scientific™ National Assembled Caps and Septa for 8-425 Standard-Opening Vials are available in a variety of colors and septum materials for a wide range of applications.
  • Open top caps can be used with any 8mm septa
  • Phenolic caps are suitable for autoclaving and low temperature applications
  • Polypropylene caps are chemically inert and suitable for most chromatography applications
  • Kim-Lok™ cap/septum assemblies feature a molded centering ring to eliminate septum movement during injection
  • PTFE/red rubber septa are economical for many GC applications
  • PTFE/silicone septa are most popular for HPLC and IC applications
  • Pre-slit septa are easier to pierce with thin-gauge needles
TRUSTED_SUSTAINABILITY
Specifications
Description PP, Black; Kim-Lok Red PTFE/White Silicone septum
Color Black
Material PTFE, Silicone, Polypropylene
Septum Kim-Lok Red PTFE/White Silicone
Type Kim-Lok Cap with Flange
Closure Size 8-425
For Use With (Equipment) 8-425 Standard Opening Vials