missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Aspartate Aminotransferase Activity Assay Kit (Fluorometric)
Shop All Bio Techne ProductsSpecifications
Specifications
| Content And Storage | Storage of components varies. See protocol for specific instructions. |
| Gene ID (Entrez) | 2805 |
| Gene Symbol | GOT1 |
| Quantity | 96 Tests |
| Synonym | aspartate aminotransferase, cytoplasmic, ASTQTL1, CASPAT, CCAT, Cysteine aminotransferase, cytoplasmic, Cysteine transaminase, cytoplasmic, EC 2.6.1.1, EC 2.6.1.3, GIG18, Glutamate oxaloacetate transaminase 1, glutamic-oxaloacetic transaminase 1, soluble (aspartate aminotransferase 1), GOT1, growth-inhibiting protein 18, Transaminase A |
| Research Category | Lipid and Metabolism |
| Species | Non-species specific |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?