Learn More
Honeywell™ Ascender ESD Safety Footwear
Click to view available options
Size:
42
Unit Size:
1 pair
Specifications
Specifications
| Certifications/Compliance | EN ISO 20345: 2011 S3 CI SRC, EN 61340-4-3:2001 |
| Liner Material | 3D |
| Material | Leather, Textile |
| Size | 42 |
| Resistant To | Water |
| Waterproof | Yes |
| Product Type | Safety Shoe |