missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ AS 2444697

Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
Description
AS 2444697 is a potent and selective interleukin-1 receptor-associated kinase 4 (IRAK4) inhibitor (IC50 = 21 nM). Displays 30-fold selectivity for IRAK4 over IRAK1. Inhibits LPS-induced TNF-α and IL-6 production in PBMCs in vitro. Renoprotective and anti-inflammatory in a rodent model of chronic kidney disease.
Specifications
Specifications
| Chemical Name or Material | N-[3-Aminocarbonyl)-1-(tetrahydro-2H-pyran-4-yl)-1H-pyrazol-4-yl]-2-(2-methyl-4-pyridinyl)-4-oxazolecarboxamide hydrochloride |
| CAS | 1287665-60-4 |
| Quantity | 10 mg |
| Target | Other Kinase Inhibitors |
| Molecular Formula | C19H20N6O4.HCl |
| Formula Weight | 432.86 |
| Purity | >98% |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction